Learn More
Sino Biological Recombinant Mouse VEGFR2/KDR Protein (ECD, His & AVI Tag), Avi-tag Biotinylated, HPLC-verified
Supplier:  Sino Biological 50998M49HB100
Encompass_Preferred
A DNA sequence encoding the Mouse KDR (P35918-1) (Met1-Glu762) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-299-2980