Learn More
American Research Products Inc Mouse Kelch like ECH associate
Supplier:  American Research Products Inc CSBEL012147MO
Encompass
Mouse Kelch-like ECH-associated protein 1 (KEAP1) ELISA kit from CUSABIO Detection Range 4 7 pg/mL-300 pg/mL Sensitivity 1 17 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27623