Learn More
Biomatik Corporation Mouse Klotho (KL) CLIA Kit
Supplier:  Biomatik Corporation EKU09979
Encompass_Preferred
Mouse Klotho (KL) CLIA Kit for detecting KL in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.15pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: O35082. Gene ID: 16591. Target Synonyms: KLa; Klotho Alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6421