Learn More
ATCC ATCC CRL-10680 - Mouse L cells expressing human CDw32 (clone 16.2 CG7)
Supplier:  ATCC CRL10680
Encompass
Mouse L cells engineered to express human CDw32 (clone 16.2 CG7). These L-cell fibroblasts are commonly used as gamma-irradiated feeder cells for B cell culture and related immunology applications. Recommended culture conditions reported in literature include DMEM supplemented with 10% fetal bovine serum and 50 μg/mL gentamicin; incubate at 37°C.
  • Stable expression of human CDw32 in an L-cell background.
  • Suitable as feeder cells when gamma-irradiated.
  • Grows in DMEM with 10% FBS.
  • Reported antibiotic supplement: 50 μg/mL gentamicin.
  • Can be cryopreserved after preparation for later use.
Catalog No. NC2593295