Learn More
American Research Products Inc Mouse Leukocyte surface antige
Supplier:  American Research Products Inc CSBEL004940MO
Encompass
Mouse Leukocyte surface antigen CD47 (CD47) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27518