Learn More
Abcam Mouse Lipocalin-2 ELISA Kit (NGAL), 1 x 96 Tests
Supplier:  Abcam AB1990831X96TESTS
Encompass_Preferred
Mouse Lipocalin-2 ELISA Kit (NGAL) is a single-wash 90-min Simplestep used to quantify Mouse Lipocalin-2 (NGAL) with a sensitivity of 1.1 pg/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10x96 plates for bulk orders - Design your own immunoassay: we also offer the conjugation-ready antibody pair - Cited in over 15 citations
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1870164