Learn More
American Research Products Inc Mouse Lipopolysaccharides LPS
Supplier:  American Research Products Inc CSBE13066M
Encompass
Mouse Lipopolysaccharides (LPS) ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26818