Learn More
BIOHIPPO Mouse LPS Microsample
SDP
Supplier:  BIOHIPPO BHE15210656
Encompass
Mouse LPS Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of LPS in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 11 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse LPS Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse LPS Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse LPS biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13855