Learn More
American Research Products Inc Mouse Macrophage mannose recep
Supplier:  American Research Products Inc CSBEL014782MO
Encompass
Mouse Macrophage mannose receptor 1 (MRC1) ELISA kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 156 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27647