Learn More
American Research Products Inc Mouse Macrophage scaVenger rec
Supplier:  American Research Products Inc CSBEL015050MO
Encompass
Mouse Macrophage scaVenger receptor types I and II (MSR1) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27373