Learn More
Sino Biological Anti-CD146/MCAM Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50794RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CD146 / MCAM (Catalog#50794-M08H; Q8R2Y2-1; Met1-Val563). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3083