Learn More
American Research Products Inc Mouse Metallothionein 2 MT 2
Supplier:  American Research Products Inc CSBE13693M
Encompass
Mouse Metallothionein-2 (MT-2) ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29411