Learn More
Encor Biotechnology Inc MOUSE MONO ANTIBODY NF-H,50UL
Supplier:  Encor Biotechnology Inc MCA9B1250UL
Encompass
Mouse Monoclonal Antibody to Neurofilament NF-HCat# MCA-9B12 MCA-9B12 is a mouse monoclonal antibody raised against native axonal phosphorylated NF-H purified from bovine spinal cord (9). MCA-9B12 recognizes the phosphorylated NF-H KSP sequences similar to other antibodies to NF-H (5,7). In some species there is some cross-reactivity with the phosphorylated KSP sequences found in the related neurofilament subunit NF-M. The antibody recognizes NF-H strongly in all mammals tested to date and also in chicken. It recognizes neurofilaments in frozen sections in tissue culture and in formalin fixed sections. We also market alternate mouse monoclonal antibodies to NF-H MCA-NAP4 and MCA-AH1 and also rabbit and chicken polyclonals RPCA-NF-H and CPCA-NF-H, all of which have similar specificities to MCA-9B12. Mouse select image at left for larger view, and on “additional info” tab for more information.
Catalog No. NC2243433