Learn More
American Research Products Inc Mouse MYO ELISA Kit
Supplier:  American Research Products Inc RDRMYOMU
Encompass
This Mouse Myoglobin (MYO) ELISA Kit is a precoated Sandwich ELISA assay designed to detect and quantify the target analyte in serum plasma tissue homogenates and other biological fluids This product is for Research Use Only
Catalog No. 50-004-57981