Learn More
American Research Products Inc Mouse N terminal pro brain nat
Supplier:  American Research Products Inc CSBE05153M
Encompass
Mouse N-terminal pro-brain natriuretic peptide NT-proBNP ELISA KIT from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-29374