Learn More
Raybiotech Inc Mouse Neprilysin ELISA Kit
Supplier:  Raybiotech Inc ELMNEPRILYSIN1
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 61 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes mouse Neprilysin.
Catalog No. 50-195-0098