Learn More
American Research Products Inc Mouse neutrophil gelatinase as
Supplier:  American Research Products Inc CSBE09410MIS
Encompass
Mouse neutrophil gelatinase-associated lipocalin NGAL ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 15 625 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26757