Learn More
Cayman Chemical Mouse ntI-HA IgG Eu-W1024 1mg
SDP
Supplier:  Cayman Chemical 165791 MG
Encompass_Preferred
Antibody: Mouse mononcolonal anti-Hemagglutinin IgG (clone 16B12) Dye: Europium Chelate W-1024 Excitation max. λ: 340 nm Emission max. λ: 615 nm Uses: TR-FRET and High throughput screening (HTS) applications
Catalog No. 50-296-5120