Learn More
Cayman Chemical Mouse ntI6 HIs IgG SureLIgh
SDP
Supplier:  Cayman Chemical 27094100 UG
Encompass_Preferred
Mouse IgG monoclonal against popular epitope tag, 6-histidine peptide conjugated to SureLight™ 488. Antibody: Anti-6His and Goat Uses: Flow cytometry/FACS and Microscopy Concentration: 100 UG/ml
Catalog No. 50-293-8537