Learn More
Cayman Chemical Mouse ntI6HIS IgG BIotIn 100ug
SDP
Supplier:  Cayman Chemical 16557100 UG
Encompass_Preferred
Mouse anti-6xHistidine IgG1 conjugated to Biotin Antibody: Mouse monoclonal anti-6xHistidine IgG, clone AD1.1.10 Binding Moiety: Biotin Uses: ELISA, microscropy, and Western blot detection Recommended dilution of 1:500-1:10,000. Starting dilution: 1:2,000)
Catalog No. 50-296-7424