Learn More
Cayman Chemical Mouse ntI6HIS IgG EuW1024 50ug
SDP
Supplier:  Cayman Chemical 1657650 UG
Encompass_Preferred
Antibody: Mouse monoclonal anti-6xHistidine IgG, clone AD1.1.10 Dye: Europium Chelate W-1024 Excitation max. λ: 340 nm Emission max. λ: 615 nm Uses: TR-FRET and High throughput screening (HTS) applications
Catalog No. 50-296-6737