Learn More
Cayman Chemical Mouse ntI6HIS IgG Unconjuga
SDP
Supplier:  Cayman Chemical 16539100 UG
Encompass_Preferred
Mouse anti-6His IgG, unconjugated Antibody: Mouse anti-6x Histidine IgG, clone AD1.1.10 Isotype: IgG1-κ Reactivity: Polyhisitidine tagged proteins (N- or C-terminus label) Uses: ELISA and Western blot analysis: 1:500-1:1,000; IHC: 1:250-750 dilution
Catalog No. 50-296-7734