Learn More
Cayman Chemical Mouse ntIcmyc IgG DyLIght 1ea
SDP
Supplier:  Cayman Chemical 112471 EA
Encompass_Preferred
Mouse Anti-c-myc IgG clone 9E10 (EQKLISEEDL) to DyLight 488 Antibody: Mouse monoclonal anti-c-myc, IgG1 Clone: 9E10 (EQKLISEEDL) Isotype: IgG1 Dye: DyLight 488 Excitation max. λ: 493 nm Emission max. λ: 518 nm Uses: Fluorescence-activated cell sorting, flow cytometry, microscopy, and protein microarrays
Catalog No. 50-295-1099