Learn More
Cayman Chemical Mouse ntIcmyc IgG DyLIght
SDP
Supplier:  Cayman Chemical 11264100 UG
Encompass_Preferred
Mouse anti-c-myc IgG clone 9E10 (EQKLISEEDL) conjugated to DyLight 650 Antibody: Mouse monoclonal Anti-C-MYC Clone: 9E10 Isotype: IgG1κ Reactivity: c-myc tagged proteins Immunogen: 9E10 epitope (EQKLISEEDL sequence) from the human c-myc protein Dye: DyLight 650 Excitation max. λ: 652 nm Emission max. λ: 672 nm Uses: Fluorescence-activated cell sorting, flow cytometry, high-throughput screening, protein microarrays, and time-resolved fluorescence energy transfer
Catalog No. 50-295-1095