Learn More
Cayman Chemical Mouse ntIcmyc IgG EuW1024 1mg
SDP
Supplier:  Cayman Chemical 165771 MG
Encompass_Preferred
Antibody: Mouse mononcolonal anti-c-myc IgG (clone 9E10) Dye: Europium Chelate W-1024 Excitation max. λ: 340 nm Emission max. λ: 615 nm Uses: TR-FRET and High throughput screening (HTS) applications Specificity: Recognizes the 9E10 epitope (EQKLISEEDL sequence) derived from the human c-myc protein.
Catalog No. 50-296-6609