Learn More
Cayman Chemical Mouse ntIDYKDDDDK IgG EuW1 1mg
SDP
Supplier:  Cayman Chemical 165781 MG
Encompass_Preferred
Antibody: Mouse mononcolonal anti-DYDDDK IgG (clone M2) Dye: Europium Chelate W-1024 Excitation max. λ: 340 nm Emission max. λ: 615 nm Uses: TR-FRET and High throughput screening (HTS) applications
Catalog No. 50-296-7748