Learn More
Cayman Chemical Mouse ntIDYKDDDDK IgG SureL
SDP
Supplier:  Cayman Chemical 27095100 UG
Encompass_Preferred
Mouse IgG monoclonal against popular epitope tag, 6-histidine peptide conjugated to SureLight™ 488.Antibody: Anti-DYKDDDDKConcentration: 100 UG/ml
Catalog No. 50-293-8319