Learn More
American Research Products Inc Mouse Ornithine carbamoyltrans
Supplier:  American Research Products Inc CSBEL017270MO
Encompass
Mouse Ornithine carbamoyltransferase mitochondrial (OTC) ELISA kit from CUSABIO Detection Range 3 12 mU/mL-200 mU/mL Sensitivity 0 78 mU/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27574