Learn More
American Research Products Inc Mouse Osteopontin OPN ELISA
Supplier:  American Research Products Inc CSBE12008M
Encompass
Mouse Osteopontin (OPN) ELISA Kit from CUSABIO Detection Range 78 pg/ml-5000 pg/ml Sensitivity 19 5 pg/ml Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-29217