Learn More
Sino Biological Mouse Progranulin Matched ELISA Antibody Pair Set 15 Plates
Supplier:  Sino Biological SEK5039615
Encompass_Preferred
The Mouse Progranulin / Granulin ELISA Pair Set is for the quantitative determination of Mouse Progranulin / Granulin.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Mouse Progranulin / Granulin coated on a 96-well plate. Standards and samples are added to the wells, and any Mouse Progranulin / Granulin present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Mouse Progranulin / Granulin monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Mouse Progranulin / Granulin present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85122