Learn More
American Research Products Inc Mouse Protein S100 B S100B E
Supplier:  American Research Products Inc CSBEL020643MO
Encompass
Mouse Protein S100-B (S100B) ELISA kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28199