Learn More
American Research Products Inc Mouse PTH ELISA Kit
Supplier:  American Research Products Inc RDRPTHMU
Encompass
This Mouse Parathyroid Hormone (PTH) ELISA Kit is a precoated Competitive Inhibition ELISA assay designed to detect and quantify the target analyte in serum plasma and other biological fluids This product is for Research Use Only
Catalog No. 50-004-59836