Learn More
Biomatik Corporation Mouse Relaxin (RLN) ELISA Kit
Supplier:  Biomatik Corporation EKU07061
Encompass_Preferred
Mouse Relaxin (RLN) ELISA Kit for detecting RLN in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 24.69-2000pg/ml. Sensitivity: 8.86pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P47932. Target Synonyms: RLXH1; Relaxin H1; Prorelaxin H1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6882