Learn More
American Research Products Inc Mouse Retinoic acid receptor r
Supplier:  American Research Products Inc CSBEL019342MO
Encompass
Mouse Retinoic acid receptor responder protein 2 (RARRES2) ELISA kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 411 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27198