Learn More
Biomatik Corporation Mouse S100 Calcium Binding Protein A16 (S100A16) ELISA Kit
Supplier:  Biomatik Corporation EKU08512
Encompass_Preferred
Mouse S100 Calcium Binding Protein A16 (S100A16) ELISA Kit for detecting S100A16 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 32pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9D708. Gene ID: 67860. Target Synonyms: S100-A16; AAG13; DT1P1A7; S100F; S100-F; Aging-associated gene 13 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8658