Learn More
American Research Products Inc Mouse secretory immunoglobulin
Supplier:  American Research Products Inc CSBE08413M
Encompass
Mouse secretory immunoglobulin A sIgA ELISA Kit from CUSABIO Detection Range 15 6 ng/mL-1000 ng/mL Sensitivity 3 9 ng/mL Sample serum plasma tissue homogenates bronchoalveolar lavage fluid Method Sandwich
Catalog No. 50-003-29110