Learn More
Bioworld Mouse Serum Albumin (MSA) (Lyophilized), 100 mg
Supplier:  Bioworld 220701041
Encompass_Preferred
Mouse Serum Albumin, Lyophilized PowderSpecification:Prepared from mouse serum Purity: ≥96%Protein level: ≥96%CAS Number: 9048-46-8 Molecular Weight: 67 kDa Derived from Webster strain mouse serum by cold alcohol fractionation.There are no heating steps involved in the preparation of this product and this product is essentially free of globulin.Usage:bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY.The product may not be used as a drug,agricultural or pesticidal product,food additive or as a household chemical.
Catalog No. 50-253-980