Learn More
Rockland Immunochemicals Mouse Serum Non-Sterile 50 mL
SDP
Supplier:  Rockland Immunochemicals D208000050
Encompass
Mouse Serum is used as a component of bioassays immunoassays or enzyme assays. Mouse Serum provides a broad spectrum of macromolecules carrier proteins for lipoid substances and trace elements attachment and spreading factors low molecular weight nutrients and hormones and growth factors that promote cell growth and health. Mouse Serum is also available as a sterile preparation for use as a cell culture supplement. Mouse Serum is ideal for investigators in Cancer Immunology and Cell Biology research. Mouse Serum can be used as a blocking agent to treat plastic surfaces membrane or tissue after they have been sensitized with primary antibody or antigen. It provides an alternative to fish gel and other blocking agents. It is effective in reducing nonspecific binding of proteins to reaction surfaces thereby maximizing signal-to-noise ratio.
Catalog No. 50-194-3965