Learn More
Sino Biological Recombinant Mouse SEZ6 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 5A0124M08H100
Encompass_Preferred
A DNA sequence encoding the Mouse SEZ6 (Q7TSK2-1) (Leu20-His922) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-2841