Learn More
Bon Opus Biosciences Recombinant Mouse SIGLEC-10 (C-mFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP106050UG
Encompass_Preferred
Recombinant Mouse SIGLEC-10 (C-mFc) is produced by Human Cells. The target gene encoding Gly17-Val546 is expressed with a C-mFc tag. Accession# Q80ZE3-1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1060
Catalog No. 50-217-2658