Learn More
Sino Biological Recombinant Mouse Siglec-10 Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 5A7659M02H1
Encompass_Preferred
A DNA sequence encoding the Mouse SIGLEC10 (Q80ZE3) (Met1-Lys543) was expressed with the fused Fc region of human IgG1 at the C-terminus.
Catalog No. 50-299-2753