Learn More
ACROBiosystems PE-Labeled Mouse SIRP alpha, His Tag (SIA-MP2H6) is produced via site-specific conjugation of PE to Mouse SIRP alpha, His Tag under optimal conditions with a proprietary technology. Mouse SIRP alpha, His Tag is expressed from human 293 cell
Supplier:  ACROBiosystems SIAMP2H625TESTS
Encompass_Preferred
Tyrosine-protein phosphatase non-receptor type substrate 1 (SHPS1) is also known as CD172 antigen-like family member A (CD172a), Macrophage fusion receptor, MyD-1 antigen, Signal-regulatory protein alpha (SIRPA or SIRP alpha) or p84, is a member of the SIRP family, and also belongs to the immunoglobulin superfamily. SIRP alpha is Ubiquitous and highly expressed in brain. SIRPA / CD172a is immunoglobulin-like cell surface receptor for CD47 and acts as docking protein and induces translocation of PTPN6, PTPN11 and other binding partners from the cytosol to the plasma membrane. SIRPA / SHPS-1 supports adhesion of cerebellar neurons, neurite outgrowth and glial cell attachment and may play a key role in intracellular signaling during synaptogenesis and in synaptic function By similarity. SIRPA / MyD1 involved in the negative regulation of receptor tyrosine kinase-coupled cellular responses induced by cell adhesion, growth factors or insulin and mediates negative regulation of phagocytosis,
Catalog No. 50-225-3978