Learn More
Sino Biological Recombinant Mouse CD98 Protein (His & AVI Tag), Avi-tag Biotinylated, HPLC-verified
Supplier:  Sino Biological 50813M56HB100
Encompass_Preferred
A DNA sequence encoding the Mouse SLC3A2 (NP_001154885.1) (Ala139-Ala565) was expressed with a N-terminal polyhistidine tag followed by an AVI tag.The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-299-3598