Learn More
American Research Products Inc Mouse soluble platelet endothe
Supplier:  American Research Products Inc CSBE08135M
Encompass
Mouse soluble platelet endothelial cell adhesion molecule 1 sPECAM-1 ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-26439