Learn More
ACROBiosystems Anti-SARS-CoV-2 Spike RBD antibody, Mouse IgG1 (9A6A11) (XBB.1.5/Omicron specific), 100 μg
Supplier: ACROBiosystems SPDY168100UG
Mouse monoclonal antibody (IgG1) that specifically recognizes the receptor-binding domain (RBD) of the SARS-CoV-2 Spike protein; reported to be specific for the XBB.1.5/Omicron lineage. Supplied lyophilized and formulated for stability; optimized for ELISA applications.
- Host species: Mouse.
- Isotype: IgG1, kappa.
- Specificity: SARS-CoV-2 Spike RBD (XBB.1.5/Omicron specific).
- Recommended application: ELISA (0.8-200 ng/mL recommended usage).
- Format: Lyophilized from 0.22 μm filtered PBS with trehalose protectant.
- Storage: Store lyophilized at -20 °C or lower; avoid repeated freeze-thaw.
- Purity: >95% by SDS-PAGE.
- Size: 100 μg (also available in 1 mg multi-vial pack).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.