Learn More
American Research Products Inc Mouse Spondin 1 SPON1 ELISA
Supplier:  American Research Products Inc CSBEL022599MO
Encompass
Mouse Spondin-1 (SPON1) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27873