Learn More
Abcam Mouse Syndecan-1 ELISA Kit, 1 x 96 tests, Sandwich ELISA, < 0.35 ng/mL sensitivity, 0.5 - 16 ng/mL detection range, Pre-coated microwell strips, Colorimetric readout, 450 nm
Supplier:  Abcam AB2731651X96TESTS
Encompass_Preferred
Mouse Syndecan-1 ELISA Kit, 1 x 96 tests, Sandwich ELISA, < 0.35 ng/mL sensitivity, 0.5 - 16 ng/mL detection range, Pre-coated microwell strips, Colorimetric readout, 450 nm
Catalog No. NC2045255