Learn More
American Research Products Inc Mouse tissue inhibitors of met
Supplier:  American Research Products Inc CSBE08004M
Encompass
Mouse tissue inhibitors of metalloproteinase 1 TIMP-1 ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 110 ng/mL Sample serum plasma cell culture supernates Method Sandwich
Catalog No. 50-003-29000