Learn More
American Research Products Inc Mouse tissue inhibitors of met
Supplier:  American Research Products Inc CSBE07387M
Encompass
Mouse tissue inhibitors of metalloproteinase 2 TIMP-2 ELISA kit from CUSABIO Detection Range 125 pg/mL-8000 pg/mL Sensitivity 31 25 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-28958