Learn More
Sino Biological Mouse TNF-α ELISpot Kit (ALP) 10 Kit (96 Tests)
Supplier:  Sino Biological EK50349A10
Encompass_Preferred
The kit applies for the detection of mouse TNF-α at the cellular level by counting TNF-α-secreting cells.
Catalog No. 50-006-96183